2-(Oxiran-2-ylmethyl)-1H-isoindole-1,3(2H)-dione 97%
Lieferant: Apollo Scientific
Synonyme:
2-(Oxiran-2-ylmethyl)isoindoline-1,3-dione, N-(2,3-Epoxypropyl)phthalimide
OR28220-100G
OR28220-25G
APOSOR28220-100GEA
169
EUR
APOSOR28220-100G
APOSOR28220-25G
2-(Oxiran-2-ylmethyl)-1H-isoindole-1,3(2H)-dione 97%
2-(Oxiran-2-ylmethyl)-1H-isoindole-1,3(2H)-dione
Formel:
C₁₁H₉NO₃ Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005896 CAS-Nummer: 5455-98-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...