3-Brom-6-chlor-2-methylpyridin

Lieferant: Apollo Scientific
Achtung

Synonyme: 5-Brom-2-chlor-6-picolin, 5-Brom-2-chlor-6-methylpyridin, 3-Brom-6-chlor-2-picolin

OR2823-100G OR2823-25G OR2823-5G
APOSOR2823-100GEA 580 EUR
APOSOR2823-100G APOSOR2823-25G APOSOR2823-5G
3-Brom-6-chlor-2-methylpyridin
3-Bromo-6-chloro-2-methylpyridine
Formel: C₆H₅BrClN
Molecular Weight: 206.47 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07375117
CAS-Nummer: 132606-40-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR