3,5-Bis(brommethyl)toluol
Lieferant: Apollo Scientific
|
Gefahr
|
OR28251-10G
OR28251-5G
OR28251-1G
APOSOR28251-10GEA
956
EUR
APOSOR28251-10G
APOSOR28251-5G
APOSOR28251-1G
3,5-Bis(brommethyl)toluol
3,5-Bis(brommethyl)toluol
Formel:
C₉H₁₀Br₂ Schmelzpunkt: 64…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00182480 CAS-Nummer: 19294-04-3 UN: 1759 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...