5-Brom-2-benzo[b]thiophencarbonsäure
Lieferant: Apollo Scientific
|
Achtung
|
OR2826-1G
OR2826-25G
APOSOR2826-1GEA
45.1
EUR
APOSOR2826-1G
APOSOR2826-25G
5-Brom-2-benzo[b]thiophencarbonsäure
5-Brom-2-benzo[b]thiophencarbonsäure
Formel:
C₉H₅BrO₂S Molecular Weight: 257,1000 g/mol Schmelzpunkt: 233…237 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD01929338 CAS-Nummer: 7312-10-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...