2-Chlor-6-hydrazinopyridin 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(6-Chloro-2-pyridyl)hydrazine
OR2830-25G
OR2830-5G
APOSOR2830-25GEA
26.1
EUR
APOSOR2830-25G
APOSOR2830-5G
2-Chlor-6-hydrazinopyridin 95%
2-Chlor-6-hydrazinopyridin
Formel:
C₅H₆ClN₃ Molecular Weight: 143.58 g/mol Schmelzpunkt: 117…120 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00214724 CAS-Nummer: 5193-03-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...