Ethyl-4-methyl-1,3-oxazol-5-carboxylat
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 4-methyloxazole-5-carboxylate
OR28307-1G
OR28307-250MG
APOSOR28307-1GEA
34.8
EUR
APOSOR28307-1G
APOSOR28307-250MG
Ethyl-4-methyl-1,3-oxazol-5-carboxylat
Ethyl-4-methyloxazole-5-carboxylate
Formel:
C₇H₉NO₃ Molecular Weight: 155.15 g/mol Schmelzpunkt: 36…38 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00062573 CAS-Nummer: 20485-39-6 EINECS: 243-848-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...