(Brommethyl)cyclopropan 98%

Lieferant: Apollo Scientific
Achtung

OR28323-100G OR28323-25G OR28323-5G
APOSOR28323-100GEA 65.4 EUR
APOSOR28323-100G APOSOR28323-25G APOSOR28323-5G
(Brommethyl)cyclopropan 98%
(Brommethyl)cyclopropan
Formel: C₄H₇Br
Molecular Weight: 135 g/mol
Siedepunkt: 101…112 °C
Dichte: 1,392 g/cm³ (25 °C)
Flammpunkt: 41 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00001306
CAS-Nummer: 7051-34-5
EINECS: 230-331-8
UN: 1993
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR