3-Pyridinecarboxamide oxime
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-Pyridylamidoxime, Nicotinamide Oxime
OR28384-100G
OR28384-25G
OR28384-5G
APOSOR28384-100GEA
39.3
EUR
APOSOR28384-100G
APOSOR28384-25G
APOSOR28384-5G
3-Pyridinecarboxamide oxime
3-Pyridinecarboxamide oxime
Formel:
C₆H₇N₃O Molecular Weight: 137.14 g/mol Schmelzpunkt: 130…135 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00265955 CAS-Nummer: 1594-58-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...