2-(4-Morpholinyl)phenol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Morpholinophenol
OR28461-10G
OR28461-1G
OR28461-250MG
APOSOR28461-10GEA
638
EUR
APOSOR28461-10G
APOSOR28461-1G
APOSOR28461-250MG
2-(4-Morpholinyl)phenol
2-(4-Morpholinyl)phenol
Formel:
C₁₀H₁₃NO₂ Molecular Weight: 179,215 g/mol Schmelzpunkt: 126…129 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00051674 CAS-Nummer: 41536-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...