5-Methyl-1,3-benzoxazole-2-thiol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Methyl-2,3-dihydro-1,3-benzoxazole-2-thione
OR28485-1G
OR28485-25G
OR28485-5G
APOSOR28485-1GEA
21.9
EUR
APOSOR28485-1G
APOSOR28485-25G
APOSOR28485-5G
5-Methyl-1,3-benzoxazole-2-thiol
5-Methyl-1,3-benzoxazole-2-thiol
Formel:
C₈H₇NOS Molecular Weight: 165,22 g/mol Schmelzpunkt: 223…225 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08447019 CAS-Nummer: 22876-22-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...