4-Chlor-2-picolin 97%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Chloro-2-methylpyridine
OR2849-25G
OR2849-5G
APOSOR2849-25GEA
66.7
EUR
APOSOR2849-25G
APOSOR2849-5G
4-Chlor-2-picolin 97%
4-Chlor-2-picolin
Formel:
C₆H₆ClN Molecular Weight: 127,57 g/mol Siedepunkt: 56 °C/15mmHg Dichte: 1,15 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00038833 CAS-Nummer: 3678-63-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...