β,4-Dinitrostyrol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-nitro-4-(2-nitrovinyl)benzene

OR28512-1G OR28512-5G
APOSOR28512-1GEA 105 EUR
APOSOR28512-1G APOSOR28512-5G
β,4-Dinitrostyrol 95%
β,4-Dinitrostyrol
Formel: C₈H₆N₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00052058
CAS-Nummer: 3156-41-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR