β,3-Dinitrostyrol

Lieferant: Apollo Scientific

Synonyme: 1-Nitro-3-(2-nitrovinyl)benzene

OR28534-10G OR28534-1G OR28534-5G
APOSOR28534-10GEA 691 EUR
APOSOR28534-10G APOSOR28534-1G APOSOR28534-5G
β,3-Dinitrostyrol
β,3-Dinitrostyrol
Formel: C₈H₆N₂O₄
Molecular Weight: 194,15 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00052063
CAS-Nummer: 882-26-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR