4,7-Dichlorisatin

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,7-Dichlor-2,3-indolindion, 4,7-Dichlor-1H-indol-2,3-dion, 4,7-Dichlor-2,3-dioxoindolin, 4,7-Dichlorindolin-2,3-dion

OR28538-1G OR28538-5G
APOSOR28538-1GEA 21.9 EUR
APOSOR28538-1G APOSOR28538-5G
4,7-Dichlorisatin
4,7-Dichlorisatin
Formel: C₈H₃Cl₂NO₂
Molecular Weight: 216,02 g/mol
Schmelzpunkt: 246…252 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047214
CAS-Nummer: 18711-13-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR