2,6-Dichlorbenzamid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2,6-Dichlorbenzoesäureamid
OR28574-100G
OR28574-25G
APOSOR28574-100GEA
174
EUR
APOSOR28574-100G
APOSOR28574-25G
2,6-Dichlorbenzamid
2,6-Dichlorbenzamid
Formel:
C₇H₅Cl₂NO Schmelzpunkt: 196…200 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007975 CAS-Nummer: 2008-58-4 EINECS: 217-918-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...