3-(2,6-Dichlorphenyl)-5-methyl-4-isoxazolcarbonsäure

Lieferant: Apollo Scientific
Achtung

OR28602-5G OR28602-10G OR28602-50G
APOSOR28602-5GEA 39.3 EUR
APOSOR28602-5G APOSOR28602-10G APOSOR28602-50G
3-(2,6-Dichlorphenyl)-5-methyl-4-isoxazolcarbonsäure
3-(2,6-Dichlorophenyl)-5-methylisoxazole-4-carboxylic acid
Formel: C₁₁H₇Cl₂NO₃
Molecular Weight: 272,09 g/mol
Schmelzpunkt: 221…225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00051658
CAS-Nummer: 3919-76-4
EINECS: 223-487-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR