tert-Butylhydrazin Hydrochlorid

Lieferant: Apollo Scientific
Achtung

OR28606-1KG OR28606-500G
APOSOR28606-1KGEA 121 EUR
APOSOR28606-1KG APOSOR28606-500G
tert-Butylhydrazin Hydrochlorid
tert-Butylhydrazin Hydrochlorid
Formel: C₄H₁₃ClN₂
Molecular Weight: 124.61 g/mol
Schmelzpunkt: 191…194 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012947
CAS-Nummer: 7400-27-3
EINECS: 231-001-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR