6-Nitro-2,3-xylidin

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dimethyl-6-nitroanilin

OR28632-25G OR28632-500G
APOSOR28632-25GEA 89.9 EUR
APOSOR28632-25G APOSOR28632-500G
6-Nitro-2,3-xylidin
6-Nitro-2,3-xylidin
Formel: C₈H₁₀N₂O₂
Molecular Weight: 166,18 g/mol
Schmelzpunkt: 115…122 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00010148
CAS-Nummer: 59146-96-2
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR