2-Amino-5-mercapto-1,3,4-thiadiazol 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Amino-1,3,4-thiadiazole-2-thiol
OR28655-500G
OR28655-100G
APOSOR28655-500GEA
79.9
EUR
APOSOR28655-500G
APOSOR28655-100G
2-Amino-5-mercapto-1,3,4-thiadiazol 99%
2-Amino-5-mercapto-1,3,4-thiadiazole
Formel:
C₂H₃N₃S₂ Schmelzpunkt: 235…243 °C (Zers.) Lagertemperatur: Kühlschrank |
MDL:
MFCD00003108 CAS-Nummer: 2349-67-9 EINECS: 219-078-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...