1H-1,2,4-Triazol-3-thiol

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Mercapto-1H-1,2,4-triazol, 3-Mercapto-1,2,4-triazol

OR28670-100G OR28670-25G OR28670-500G
APOSOR28670-100GEA 29 EUR
APOSOR28670-100G APOSOR28670-25G APOSOR28670-500G
1H-1,2,4-Triazol-3-thiol
1H-1,2,4-Triazol-3-thiol
Formel: C₂H₃N₃S
Molecular Weight: 101,13 g/mol
Schmelzpunkt: 217…224 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00005229
CAS-Nummer: 3179-31-5
EINECS: 221-656-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR