Isonipecotamid 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Piperidine-4-carboxamide

OR28687-100G OR28687-250G OR28687-25G
APOSOR28687-100GEA 36.4 EUR
APOSOR28687-100G APOSOR28687-250G APOSOR28687-25G
Isonipecotamid 98%
Isonipecotamid
Formel: C₆H₁₂N₂O
Molecular Weight: 128.18 g/mol
Schmelzpunkt: 145…149 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038012
CAS-Nummer: 39546-32-2
EINECS: 254-501-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR