4-Isopropylbenzonitril

Lieferant: Apollo Scientific
Achtung

OR28694-5G
APOSOR28694-5GEA 132 EUR
APOSOR28694-5G
4-Isopropylbenzonitril
4-Isopropylbenzonitril
Formel: C₁₀H₁₁N
Molecular Weight: 145,2 g/mol
Siedepunkt: 88…90 °C/5mmHg
Lagertemperatur: Raumtemperatur
MDL: MFCD00052966
CAS-Nummer: 13816-33-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR