6-Nitroveratrumaldehyd 96%
Lieferant: Apollo Scientific
Synonyme:
4,5-Dimethoxy-2-nitrobenzaldehyd, 3,4-Dimethoxy-6-nitrobenzaldehyd
OR28696-100G
APOSOR28696-100GEA
145
EUR
APOSOR28696-100G
6-Nitroveratrumaldehyd 96%
6-Nitroveratrumaldehyd
Formel:
C₉H₉NO₅ Molecular Weight: 211,17 g/mol Schmelzpunkt: 132…133 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00007134 CAS-Nummer: 20357-25-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...