3-tert-Butylbenzoic acid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(tert-Butyl)benzoic acid
OR2870-5G
APOSOR2870-5GEA
248
EUR
APOSOR2870-5G
3-tert-Butylbenzoic acid
3-tert-Butylbenzoic acid
Formel:
C₁₁H₁₄O₂ Molecular Weight: 178.23 g/mol Schmelzpunkt: 129…130 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01463861 CAS-Nummer: 7498-54-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...