1-(3-Methyl-2-pyrazinyl)ethanon
Lieferant: Apollo Scientific
Synonyme:
1-(3-Methylpyrazinyl)ethan-1-on
OR28702-100G
OR28702-25G
APOSOR28702-100GEA
368
EUR
APOSOR28702-100G
APOSOR28702-25G
1-(3-Methyl-2-pyrazinyl)ethanon
2-Acetyl-3-methylpyrazine
Formel:
C₇H₈N₂O Molecular Weight: 136,15 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014612 CAS-Nummer: 23787-80-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...