Ethyl-4,6-dihydroxynicotinate

Lieferant: Apollo Scientific
Gefahr

Synonyme: Ethyl 4,6-dihydroxynicotinate

OR2871-1G OR2871-5G
APOSOR2871-1GEA 21.9 EUR
APOSOR2871-1G APOSOR2871-5G
Ethyl-4,6-dihydroxynicotinate
Ethyl-4,6-dihydroxynicotinate
Formel: C₈H₉NO₄
Molecular Weight: 183,17 g/mol
Schmelzpunkt: 214…217 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00834976
CAS-Nummer: 6975-44-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR