Dimethyl-2-aminoterephthalat

Lieferant: Apollo Scientific
Achtung

Synonyme: Dimethyl aminoterephthalate, 2-Aminodimethyl terephthalate

OR28713-5G
APOSOR28713-5GEA 21.9 EUR
APOSOR28713-5G
Dimethyl-2-aminoterephthalat
Dimethyl-2-aminoterephthalat
Formel: C₁₀H₁₁NO₄
Molecular Weight: 209.2 g/mol
Schmelzpunkt: 127…134 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00008427
CAS-Nummer: 5372-81-6
EINECS: 226-364-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR