4-Methoxybenzyl isocyanate 98%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-Methoxybenzylisocyanate

OR2873-1G
APOSOR2873-1GEA 0 EUR
APOSOR2873-1G
4-Methoxybenzyl isocyanate 98%
4-Methoxybenzyl isocyanate

  • May form small amount of sediment.

Formel: C₉H₉NO₂
Molecular Weight: 163,176 g/mol
Siedepunkt: 60…66 °C/0,6mmHg
Dichte: 1,143 g/cm³ (25 °C)
Flammpunkt: 113 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00673064
CAS-Nummer: 56651-60-6
UN: 2206
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR