Ethyl-2-chlor-4-oxazolcarboxylat 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Ethyl 2-chlorooxazole-4-carboxylate
OR2874-1G
OR2874-5G
OR2874-25G
APOSOR2874-1GEA
21.9
EUR
APOSOR2874-1G
APOSOR2874-5G
APOSOR2874-25G
Ethyl-2-chlor-4-oxazolcarboxylat 95%
Ethyl-2-chlor-4-oxazolcarboxylat
Formel:
C₆H₆ClNO₃ Molecular Weight: 175,57 g/mol Schmelzpunkt: 97…98 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD06660120 CAS-Nummer: 460081-18-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...