5-Bromuracil 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Bromuracil, 5-Brom-2,4-dihydroxypyrimidin
OR28754-500G
APOSOR28754-500GEA
142
EUR
APOSOR28754-500G
5-Bromuracil 98%
5-Bromuracil
Formel:
C₄H₃BrN₂O₂ Molecular Weight: 190,98 g/mol Schmelzpunkt: >300 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006017 CAS-Nummer: 51-20-7 EINECS: 200-084-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...