1-Adamantancarbonsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 1-Adamantanecarboxylic acid, Tricyclo[3.3.1.1(3,7)]decane-1-carboxylic acid

OR28762-100G OR28762-25G OR28762-500G
APOSOR28762-100GEA 40.6 EUR
APOSOR28762-100G APOSOR28762-25G APOSOR28762-500G
1-Adamantancarbonsäure 98%
1-Adamantancarbonsäure
Formel: C₁₁H₁₆O₂
Molecular Weight: 180.25 g/mol
Schmelzpunkt: 172…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00074720
CAS-Nummer: 828-51-3
EINECS: 212-584-6

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR