2-Ethyl-4-methyl-1H-imidazole-5-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
OR28797-10G
OR28797-1G
OR28797-5G
APOSOR28797-10GEA
136
EUR
APOSOR28797-10G
APOSOR28797-1G
APOSOR28797-5G
2-Ethyl-4-methyl-1H-imidazole-5-carbaldehyde
2-Ethyl-4-methyl-1H-imidazole-5-carbaldehyde
Formel:
C₇H₁₀N₂O Molecular Weight: 138,17 g/mol Schmelzpunkt: 104…105 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173727 CAS-Nummer: 88634-80-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...