4-Methyl-1H-imidazole-5-carbaldehyde
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Methyl-5-imidazolecarboxaldehyde
OR28798-5G
OR28798-10G
OR28798-1G
APOSOR28798-5GEA
46.4
EUR
APOSOR28798-5G
APOSOR28798-10G
APOSOR28798-1G
4-Methyl-1H-imidazole-5-carbaldehyde
4-Methyl-1H-imidazole-5-carbaldehyde
Formel:
C₅H₆N₂O Molecular Weight: 110,12 g/mol Schmelzpunkt: 166…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173728 CAS-Nummer: 68282-53-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...