2-Phenyl-1H-imidazole-4-carbaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR28799-1G
OR28799-5G
APOSOR28799-1GEA
85.7
EUR
APOSOR28799-1G
APOSOR28799-5G
2-Phenyl-1H-imidazole-4-carbaldehyde 95%
2-Phenyl-1H-imidazole-4-carbaldehyde
Formel:
C₁₀H₈N₂O Molecular Weight: 172.19 g/mol Schmelzpunkt: 168…170 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00173729 CAS-Nummer: 68282-47-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...