6-Aminopenicillansäure 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
(2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 6-APA 6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid 6-amino-2,2-dimethyl-5-oxoperhydroazeto[2,1-b][1,3]thiazole-3-carboxylic acid, (2S,5R,6R)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, (+)-6-Aminopenicillanic acid, (2S,5R,6R)-(+)-6-Amino-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
OR28860-500G
APOSOR28860-500GEA
97.3
EUR
APOSOR28860-500G
6-Aminopenicillansäure 97%
6-Aminopenicillanic acid
Formel:
C₈H₁₂N₂O₃S Molecular Weight: 216,26 g/mol Schmelzpunkt: 198…200 °C (Zersetzung) Lagertemperatur: Kühlschrank |
MDL:
MFCD00005176 CAS-Nummer: 551-16-6 EINECS: 208-993-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...