6-Methoxyanthranilsäure 97%

Lieferant: Apollo Scientific
Achtung

OR2887-100G OR2887-25G
APOSOR2887-100GEA 352 EUR
APOSOR2887-100G APOSOR2887-25G
6-Methoxyanthranilsäure 97%
6-Methoxyanthranilsäure
Formel: C₈H₉NO₃
Molecular Weight: 167.16 g/mol
Schmelzpunkt: 83…86 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD01941328
CAS-Nummer: 53600-33-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR