Arecoliniumbromid

Lieferant: Apollo Scientific
Gefahr

Synonyme: Arecoline hydrobromide

OR28886-5G OR28886-25G OR28886-100G
APOSOR28886-5GEA 58 EUR
APOSOR28886-5G APOSOR28886-25G APOSOR28886-100G
Arecoliniumbromid
Arecoliniumbromid
Formel: C₈H₁₄BrNO₂
Molecular Weight: 236.11 g/mol
Schmelzpunkt: 171…176 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00039041
CAS-Nummer: 300-08-3
EINECS: 206-087-3
UN: 1544
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR