4-Isopropyl-3-methylaniline hydrochloride 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Isopropyl-m-toluidiniumchlorid

OR28929-1G OR28929-5G
APOSOR28929-1GEA 29 EUR
APOSOR28929-1G APOSOR28929-5G
4-Isopropyl-3-methylaniline hydrochloride 95%
4-Isopropyl-3-methylaniline hydrochloride
Formel: C₁₀H₁₆ClN
Molecular Weight: 185,70 g/mol
Schmelzpunkt: 222…225 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00156562
CAS-Nummer: 4534-11-6
EINECS: 224-876-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR