4-(1,2,3-Thiadiazol-4-yl)phenol 95+%
Lieferant: Apollo Scientific
|
Achtung
|
OR29038-1G
OR29038-250MG
APOSOR29038-1GEA
81.2
EUR
APOSOR29038-1G
APOSOR29038-250MG
4-(1,2,3-Thiadiazol-4-yl)phenol 95+%
4-(1,2,3-Thiadiazol-4-yl)phenol
Formel:
C₈H₆N₂OS Siedepunkt: 105 °C/0,75mmHg Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052106 CAS-Nummer: 59834-05-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...