5-Oxo-2,3-dihydro-5H-[1,3]thiazolo[3,2-a]pyrimidine-6-carboxylic acid 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,3-Dihydro-5-oxo-5H-[1,3]thiazolo[3,2-a]pyrimidine-6-carboxylic acid

OR29064-1G OR29064-250MG
APOSOR29064-1GEA 130 EUR
APOSOR29064-1G APOSOR29064-250MG
5-Oxo-2,3-dihydro-5H-[1,3]thiazolo[3,2-a]pyrimidine-6-carboxylic acid 95%
5-Oxo-2,3-dihydro-5H-[1,3]thiazolo[3,2-a]pyrimidine-6-carboxylic acid
Formel: C₇H₆N₂O₃S
Molecular Weight: 198,2 g/mol
Schmelzpunkt: 235…238 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00085018
CAS-Nummer: 32084-55-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR