3-Chloro-4-hydrazinobenzonitrile
Lieferant: Apollo Scientific
|
Achtung
|
OR29065-1G
OR29065-250MG
APOSOR29065-1GEA
127
EUR
APOSOR29065-1G
APOSOR29065-250MG
3-Chloro-4-hydrazinobenzonitrile
3-Chloro-4-hydrazinobenzonitrile
Formel:
C₇H₆ClN₃ Molecular Weight: 167.6 g/mol Schmelzpunkt: 125…126 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01571093 CAS-Nummer: 254880-25-6 UN: 2811 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...