2-Amino-3,5-dibrom-4-methylpyridin
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3,5-Dibromo-4-methylpyridin-2-amine
OR2908-25G
OR2908-5G
APOSOR2908-25GEA
24.8
EUR
APOSOR2908-25G
APOSOR2908-5G
2-Amino-3,5-dibrom-4-methylpyridin
2-Amino-3,5-dibromo-4-methylpyridine
Formel:
C₅H₄Br₂N₂ Molecular Weight: 265,9300 g/mol Schmelzpunkt: 123…127 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01646065 CAS-Nummer: 3430-29-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...