2-Chloro-1H-indole-3-carbaldehyde

Lieferant: Apollo Scientific
Achtung

OR29092-1G OR29092-5G
APOSOR29092-1GEA 29 EUR
APOSOR29092-1G APOSOR29092-5G
2-Chloro-1H-indole-3-carbaldehyde
2-Chloro-1H-indole-3-carbaldehyde
Formel: C₉H₆ClNO
Molecular Weight: 179.61 g/mol
Schmelzpunkt: 211…212 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00111620
CAS-Nummer: 5059-30-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR