3-Brom-2-chlor-5-thiophensulfonylchlorid
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
4-Bromo-5-chlorothiophene-2-sulphonyl chloride
OR29142-1G
OR29142-5G
OR29142-25G
APOSOR29142-1GEA
46.4
EUR
APOSOR29142-1G
APOSOR29142-5G
APOSOR29142-25G
3-Brom-2-chlor-5-thiophensulfonylchlorid
3-Brom-2-chlor-5-thiophensulfonylchlorid
Formel:
C₄HBrCl₂O₂S₂ Molecular Weight: 295,9895 g/mol Schmelzpunkt: 63…65 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00052094 CAS-Nummer: 166964-35-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...