5-Brom-2-chlor-4-methyl-3-nitropyridin 95%

Lieferant: Apollo Scientific
Achtung

OR2915-1G OR2915-5G OR2915-10G
APOSOR2915-1GEA 50.9 EUR
APOSOR2915-1G APOSOR2915-5G APOSOR2915-10G
5-Brom-2-chlor-4-methyl-3-nitropyridin 95%
5-Brom-2-chlor-4-methyl-3-nitropyridin
Formel: C₆H₄BrClN₂O₂
Molecular Weight: 251,47 g/mol
Schmelzpunkt: 68…70 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD06659505
CAS-Nummer: 884495-15-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR