Pyrazine-2-carbothioamide

Lieferant: Apollo Scientific
Gefahr

OR29190-1G OR29190-250MG OR29190-5G
APOSOR29190-1GEA 145 EUR
APOSOR29190-1G APOSOR29190-250MG APOSOR29190-5G
Pyrazine-2-carbothioamide
Pyrazine-2-carbothioamide
Formel: C₅H₅N₃S
Molecular Weight: 139,18 g/mol
Schmelzpunkt: 198…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00173663
CAS-Nummer: 4604-72-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR