4-Chlor-4'-nitrobenzophenon

Lieferant: Apollo Scientific

Synonyme: (4-Chlorophenyl)(4-nitrophenyl)methanone

OR2920-1G OR2920-5G
APOSOR2920-1GEA 69.6 EUR
APOSOR2920-1G APOSOR2920-5G
4-Chlor-4'-nitrobenzophenon
4-Chlor-4'-nitrobenzophenon
Formel: C₁₃H₈ClNO₃
Molecular Weight: 261.6605 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00047743
CAS-Nummer: 7497-60-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR