Cyclohexyl(phenyl)methanol

Lieferant: Apollo Scientific
Achtung

OR29308-1G OR29308-250MG
APOSOR29308-1GEA 65.4 EUR
APOSOR29308-1G APOSOR29308-250MG
Cyclohexyl(phenyl)methanol
Cyclohexyl(phenyl)methanol
Formel: C₁₃H₁₈O
Lagertemperatur: Raumtemperatur
MDL: MFCD00019357
CAS-Nummer: 945-49-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR