4-Cyan-2-methylanilin
Lieferant: Apollo Scientific
Synonyme:
4-Cyano-2-methylaniline
OR2931-25G
APOSOR2931-25GEA
203
EUR
APOSOR2931-25G
4-Cyan-2-methylanilin
4-Cyan-2-methylanilin
Formel:
C₈H₈N₂ Molecular Weight: 132,16 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093969 CAS-Nummer: 78881-21-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...