2-Amino-3,5-dimethylbenzoesäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,5-Dimethylanthranilic acid

OR2932-10G OR2932-25G OR2932-5G
APOSOR2932-10GEA 94.4 EUR
APOSOR2932-10G APOSOR2932-25G APOSOR2932-5G
2-Amino-3,5-dimethylbenzoesäure 97%
2-Amino-3,5-dimethylbenzoic acid
Formel: C₉H₁₁NO₂
Molecular Weight: 165,19 g/mol
Schmelzpunkt: 189…196 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00017099
CAS-Nummer: 14438-32-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR